Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Antibody (OTI1A6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274332
Description
Sphingosine Kinase 1/SPHK1 Monoclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:400-4000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| Mouse | |
| 43.8 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1A6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| SPHK1 | |
| Full length human recombinant protein of human SPHK1(NP_068807) produced in HEK293T cell. | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 8877 | |
| Human, Monkey | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction