Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sphingosine Kinase 2/SPHK2 Antibody (9C5E1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$560.50
Specifications
Antigen | Sphingosine Kinase 2/SPHK2 |
---|---|
Clone | 9C5E1 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Description
Sphingosine Kinase 2/SPHK2 Monoclonal specifically detects Sphingosine Kinase 2/SPHK2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Sphingosine Kinase 2/SPHK2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Apoptosis | |
PBS with 0.05% Sodium Azide | |
56848 | |
Synthesized peptide of human SK2 (AA: 36-52). | |
Primary | |
69.2 kDa |
9C5E1 | |
Western Blot, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2 | |
SPHK2 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title