Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sphingosine Kinase 2/SPHK2, p Thr578 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323393
Description
Sphingosine Kinase 2/SPHK2 Polyclonal antibody specifically detects Sphingosine Kinase 2/SPHK2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Sphingosine Kinase 2/SPHK2 | |
Polyclonal | |
Western Blot 1:500 | |
EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2 | |
Phospho-Sphingosine Kinase 2/SPHK2 (Thr-578) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Thr-578 in human Sphingosine Kinase 2/SPHK2. This peptide sequence is highly conserved in rat and mouse Sphingosine Kinase 2/SPHK2 proteins, and is not well conserved in SK1. [UniProt# Q9NRA0] | |
0.1 mL | |
Apoptosis | |
56848 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction