Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPHK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587359 Supplier Invitrogen™ Supplier No. PA587359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLMLTERRNH ARELVRSEEL GRWDALVVMS GDGLMHEVVN GLMERPDWET AIQKPLCSLP AGSGNALAAS LNHYAGYEQV TNEDLLTNCT LLLCRRLLSP MNLLSLHTAS GLRLFSVLSL AWGFIADVDL ESEKYRRLGE MRFTLGTFLR LAALRTYRGR LAYLPVGRVG SKTPASPVVV QQGPVDAHLV PLEEPVPSHW TVVPDEDFVL VLALLHSHLG SEMFAAPMGR CAAGVMHLFY VRAGVSRAML LRLFLAMEKG RHMEYECPYL VYVPVVAFRL EPKDGKGVFA VDGELMVSEA VQGQVHPNYF WMVSGCVEPP PSWKPQQMPP PEEPL; Positive Samples: SW480, HeLa, Mouse kidney; Cellular Location: Cell membrane, Cytoplasm, Nucleus.

SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signaling molecule sphingosine-1-phosphate (S1P). S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapeutics for the treatment of cancer. SPHK1 catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPHK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPHK1
Gene Accession No. Q8CI15, Q9NYA1
Gene Alias 1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1
Gene Symbols SPHK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-384 of human SPHK1 (NP_0011360741).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20698, 8877
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.