Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPHK1 Polyclonal Antibody
SPHK1 Polyclonal Antibody
Supplier: Thermo Scientific PA514068
Description
SPHK1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
SPHK1 | |
Polyclonal | |
Unconjugated | |
SPHK1 | |
1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
170897, 20698, 8877 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8CI15, Q91V26, Q9NYA1 | |
SPHK1 | |
KLH conjugated synthetic peptide between 286-315 amino acids from the central region of human SPHK1. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction