Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPHK2 Polyclonal Antibody, Abbomax

Catalog No. 502203999
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3999 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3999 Supplier Abbomax Inc Supplier No. 601660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HELA cells Cellular location: Cytoplasmic.

SPHK2 Polyclonal antibody specifically detects SPHK2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SPHK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Sphk2
Gene Accession No. Q9NRA0
Gene Alias C76851; SK 2; SK2; SK-2; Sphingosine kinase 2; sphingosine kinase type 2; Sphk2; SPK 2; SPK-2; Spk2, Sk2
Host Species Rabbit
Immunogen A full length of SPHK2 recombinant protein was used as an immunogen
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.