Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPHKAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561675 Supplier Invitrogen™ Supplier No. PA561675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQSTLQTKHP DIYCITDFAE ELADTVVSMA TEIAAICLDN SSGKQPWFCA WKRGSEFLMT PNVPCRSLKR KKESQGSGTA VRKHKPPRLS EIKRKTDEHP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.

Anchoring protein that binds preferentially to the type I regulatory subunit of c-AMP-dependent protein kinase (PKA type I) and targets it to distinct subcellular compartments. May act as a converging factor linking cAMP and sphingosine signaling pathways. Plays a regulatory role in the modulation of SPHK1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPHKAP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPHKAP
Gene Accession No. Q2M3C7
Gene Alias A-kinase anchor protein SPHKAP; KIAA1678; SKIP; Sphingosine kinase type 1-interacting protein; SPHK1 (sphingosine kinase type 1) interacting protein; SPHK1 interactor, AKAP domain containing; SPHK1-interactor and AKAP domain-containing protein; SPHKAP
Gene Symbols SPHKAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPHKAP. Recombinant protein control fragment (Product #RP-99658).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.