Learn More
Invitrogen™ SPHKAP Polyclonal Antibody
Description
Recommended positive controls: NT2D1, IMR32, MCF-7, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (93%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SPHKAP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SPHKAP |
| Gene Accession No. | Q2M3C7 |
| Gene Alias | A-kinase anchor protein SPHKAP; KIAA1678; SKIP; Sphingosine kinase type 1-interacting protein; SPHK1 (sphingosine kinase type 1) interacting protein; SPHK1 interactor, AKAP domain containing; SPHK1-interactor and AKAP domain-containing protein; SPHKAP |
| Gene Symbols | SPHKAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.