Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spi-B Antibody, DyLight 350 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298958UV
Description
Spi-B Polyclonal antibody specifically detects Spi-B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Spi-B | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
6689 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Spi-B. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction