Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spi2A Antibody (MoFo 29.2), Novus Biologicals™
SDP

Catalog No. NB10074541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074541 Supplier Novus Biologicals Supplier No. NB10074541
Only null left
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Spi2A Monoclonal specifically detects Spi2A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Spi2A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MoFo 29.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500
Formulation PBS with 1mg/ml BSA with 0.05% Sodium Azide
Gene Alias Serine protease inhibitor 2A, Serine protease inhibitor A3G, Serpin 2A, Serpin A3G, Serpina3g
Gene Symbols Serpina3g
Host Species Armenian Hamster
Immunogen Synthetic peptide corresponding to residues N(418) P E R S M N F P N G E G A S S Q R L C(436) of hamster Spi2A protein.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20715
Test Specificity Spi2A (MoFo 29.2)
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.