Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPIB Monoclonal Antibody (4G5)
GREENER_CHOICE

Catalog No. PIMA517179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517179 Supplier Invitrogen™ Supplier No. MA517179
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17179 targets SPIB in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17179 immunogen is purified recombinant fragment of human SPIB (amino acids: 200-252) expressed in E. Coli. MA5-17179 detects SPIB which has a predicted molecular weight of approximately 28.8kDa.

Spi-B is a transcription factor belonging to a subfamily of ETS transcription factors with a conserved ETS domain that mediates specific DNA binding. Spi-B may form homotypic interactions and is known to interact with IRF4, CREBBP, EP300, SPI1/PU.1 related, JUN and TBP. As a sequence specific transcriptional activator it binds to the PU-box, a purine-rich DNA sequence (5'-GAGGAA-3') that can act as a lymphoid-specific enhancer. Two isoforms of Spi-B are reported. Spi-B is usually expressed in plasmacytoid dendritic cells (pDCs), also known as type 2 DC precursors (pre-DC2) or natural interferon (IFN)-producing cells and promotes their development and block viral replication. Spi-B is not expressed in T-cells or granulocytes but may be present in stem cell populations of the liver and B-cells where it helps in B-cell receptor (BCR) signaling, normal B-cell development and antigenic stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIB
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4G5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene SPIB
Gene Accession No. O35906, Q01892
Gene Alias B42b-like; Ets transcription factor Spi-B; Spi-1/PU.1 related; Spib; SPI-B; Spi-B transcription factor; Spi-B transcription factor (Spi-1/PU.1 related); Transcription factor Spi-B
Gene Symbols SPIB
Host Species Mouse
Immunogen Purified recombinant fragment of human SPIB (amino acids: 200-252) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 272382, 6689
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
What was the pH of the buffer used for validating the SPIB Monoclonal Antibody (4G5) (Cat. No. MA5-17179)?

The pH of the buffer used for validating the SPIB Monoclonal Antibody (Cat. No. MA5-17179) was pH 6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.