Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPIB Mouse anti-Human, Mouse, Clone:4G5, Abnova™
Mouse monoclonal antibody raised against recombinant human SPIB.
Supplier: Abnova Corporation MAB17740
Description
SPI1 (MIM 165170) and SPIB are members of a subfamily of ETS (see ETS1; MIM 164720) transcription factors. ETS proteins share a conserved ETS domain that mediates specific DNA binding. SPIB and SPI1 bind to a purine-rich sequence, the PU box (5-prime-GAGGAA-3-prime).[supplied by OMIM]Specifications
SPIB | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry (1:200-1:1000) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
SPIB | |
Mouse | |
100 μg | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
4G5 | |
Spi-B transcription factor (Spi-1/PU.1 related) | |
In ascites (0.03% sodium azide) | |
SPI-B | |
Recombinant protein corresponding to amino acids 200-252 of human SPIB from E. coli. | |
6689 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction