Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 157681AP150UL
Description
Spi-B is a transcription factor belonging to a subfamily of ETS transcription factors with a conserved ETS domain that mediates specific DNA binding. Spi-B may form homotypic interactions and is known to interact with IRF4, CREBBP, EP300, SPI1/PU.1 related, JUN and TBP. As a sequence specific transcriptional activator it binds to the PU-box, a purine-rich DNA sequence (5′-GAGGAA-3′) that can act as a lymphoid-specific enhancer. Two isoforms of Spi-B are reported. Spi-B is usually expressed in plasmacytoid dendritic cells (pDCs), also known as type 2 DC precursors (pre-DC2) or natural interferon (IFN)-producing cells and promotes their development and block viral replication. Spi-B is not expressed in T-cells or granulocytes but may be present in stem cell populations of the liver and B-cells where it helps in B-cell receptor (BCR) signaling, normal B-cell development and antigenic stimulation.Specifications
| SPIB | |
| Polyclonal | |
| Unconjugated | |
| SPIB | |
| SPI B, SPIB, Transcription factor Spi B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 272382, 499146, 6689 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35906, Q01892, Q5EBA3 | |
| Spib | |
| SPIB Fusion Protein Ag8404 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction