Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPIC Mouse anti-Human, Clone: PCRP-SPIC-2C5, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen SPIC
Clone PCRP-SPIC-2C5
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122614
SDP
View Documents
Novus Biologicals
NBP30757020UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

SPIC Monoclonal specifically detects SPIC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications

Specifications

SPIC
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Unconjugated
Mouse
Human
MGC40611, SPI-C, Spi-C transcription factor (Spi-1/PU.1 related), transcription factor Spi-C
Recombinant full-length human SPIC protein
Primary
Store at 4°C.
PCRP-SPIC-2C5
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
121599
IgG2b
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.