Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPICE Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564713
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4713 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4713 Supplier Bethyl Laboratories, Inc Supplier No. A303273AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan

Regulator required for centriole duplication, for proper bipolar spindle formation and chromosome congression in mitosis.

Specifications

Antigen SPICE
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SPICE1
Gene Accession No. Q8N0Z3
Gene Alias CCDC52, SPICE, coiled-coil domain containing 52, coiled-coil domain-containing protein 52, spindle and centriole protein, spindle and centriole-associated protein 1
Gene Symbols SPICE1
Host Species Rabbit
Immunogen Between 805 and 855
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152185
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.