Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPICE1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297999PCP
Description
SPICE1 Polyclonal antibody specifically detects SPICE1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SPICE1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CCDC52, FLJ26064, SPICE, spindle and centriole associated protein 1 | |
Produced in rabbits immunized with E. coli-derived Human SPICE1 fragment. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication | |
152185 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction