Learn More
Invitrogen™ SPICE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121147
Description
Positive test controls include: 293T, HeLa, BT-474, MCF7. The target is usually found in the following locations: Cytoplasm, centriole, centrosome, cytoskeleton, microtubule organizing center, spindle. Immunogen sequence: MSFVRVNRCG PRVGVRKTPK VKKKKTSVKQ EWDNTVTDLT VHRATPEDLV RRHEIHKSKN RALVHWELQE KALKRKWRKQ KPETLNLEKR RLSIMKEILS DQYQMQDVLE KSDHLIAAAK ELFPRRRTGF PNVTVAPDSS QGPIVVNQDP ITQSIFNESV IEPQALNDVD GEEEGTVNSQ SGESENENEL DNSLNSQSNT NTDRFLQQLT EENFELISKL.
Regulator required for centriole duplication, for proper bipolar spindle formation and chromosome congression in mitosis.
Specifications
| SPICE1 | |
| Polyclonal | |
| Unconjugated | |
| SPICE1 | |
| CCDC52; coiled-coil domain containing 52; coiled-coil domain-containing protein 52; D16Ertd480e; RGD1310729; SPICE; SPICE1; spindle and centriole associated protein 1; spindle and centriole protein; Spindle and centriole-associated protein; Spindle and centriole-associated protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 152185, 212514, 288111 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.78 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5RKG1, Q8C804, Q8N0Z3 | |
| SPICE1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human SPICE1 (NP_653319.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.