Learn More
Dynalon™ HDPE, Spigot
SCI_ED
Catalog No. S29269 Shop All Dynalab Corporation Products
This item is not returnable. View return policy
Catalog No. S29269 Supplier Dynalon™ Supplier No. 212905/EMD
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Use With: HDPE Heavy Walled Carboys
Plastic 3/4 in. spigot with gasket is leakproof and screws directly into a welded wall insert on the HDPE heavy walled carboys.
Specifications
Autoclavable Not Autoclavable
Description Spigot HDPE for Carboy No. 14-400-100, -101, -102, -103; 3/4 in.
For Use With (Application) Carboy
For Use With (Equipment) HDPE Heavy Walled Carboys
Includes Spigot
Material HDPE
Max. Temperature (English) 248°F
Max. Temperature (Metric) 120°C
Product Type Spigot