Learn More
Spike RBD Antibody (1038345), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB107073MFV610
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in MERS-CoV samples. It is validated for Flow CytometrySpecifications
Spike RBD | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Chinese Hamster Ovary cell line CHO-derived MERS-CoV Spike RBD, Glu367-Tyr606, Accession # YP_007188579.1 | |
RUO | |
MERS-CoV | |
IgG2a |
Flow Cytometry | |
1038345 | |
Flow Cytometry | |
Mouse | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |