Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spike RBD Antibody (1042425), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10827P
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
Spike RBD | |
Monoclonal | |
PE | |
PBS | |
Chinese Hamster Ovary cell line CHO-derived SARS-CoV Spike RBD, Arg306-Phe527, Accession # NP_0828851.1 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, Immunocytochemistry | |
1042425 | |
Western Blot, Flow Cytometry, Immunocytochemistry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction