Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spike RBD Antibody (1049349), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB11055E
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
Spike RBD | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD, Val16-Pro681, Accession # YP_009724390.1 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry, Immunocytochemistry | |
1049349 | |
Flow Cytometry, Immunocytochemistry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction