Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spike RBD Antibody (2847A), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB11198W
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot, Flow CytometrySpecifications
Spike RBD | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
CHO-derived sars-cov-2 Spike RBD protein, Arg319-Phe541, Accession # YP_009724390.1 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry | |
2847A | |
Western Blot, Flow Cytometry | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction