Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike RBD LlaMABodyTM VHH His-tag Antibody, R&D Systems™

Llama Monoclonal Antibody
Supplier: R&D Systems LMAB12503SP
This item is not returnable.
View return policy
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Blocking AssaySpecifications
Spike RBD | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Llama | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Blocking Assay | |
L009.2.69N | |
Blockade of Receptor-ligand Interaction | |
YP_009724390.1 | |
Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD protein, Arg319-Phe541, Accession # YP_009724390.1 | |
25 μg | |
Primary | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction