Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MERS-CoV Spike RBD MAb (Cl 1038342), R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $420.50
Specifications
Antigen | Spike RBD |
---|---|
Clone | 1038342 |
Dilution | Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL |
Applications | Blocking Assay, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB107071SP
![]() |
R&D Systems
MAB107071SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10707110
![]() |
R&D Systems
MAB107071100 |
100 μg |
Each for $420.50
|
|
|||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in MERS-CoV samples. It is validated for Blocking Assay, ImmunocytochemistrySpecifications
Spike RBD | |
Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Chinese Hamster Ovary cell line CHO-derived MERS-CoV Spike RBD, Glu367-Tyr606, Accession # YP_007188579.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1038342 | |
Blocking Assay, Immunocytochemistry | |
Unconjugated | |
Mouse | |
MERS-CoV | |
YP_007188579.1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title