Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spike S1 Rabbit, anti-SARS-CoV-2, Unconjugated, MP Biomedicals™

Catalog No. MP08L100003 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100003 Supplier MP Biomedicals, Inc Supplier No. 08L100003
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SARS-CoV-2 Spike S1 Rabbit Monoclonal Antibody has been experimentally validated in ELISA, WB, IF assays.

  • The spike protein (S) of coronavirus (CoV) attaches the virus to its cellular receptor, angiotensin-converting enzyme 2 (ACE2)
  • The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity
  • Offers high specificity and potency against SARS-CoV-2 antigens in immunological and biochemical assays and consistency between lots
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spike S1
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Dilution ELISA: 1:1000 to 1:5000; Flow Cytometry: 1:50 to 1:200; Immunofluorescence: 1:50 to 1:200; Immunoprecipitation: 1:50 to 1:200
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol, pH7.3 to 7.4.
Gene Alias S protein; Spike glycoprotein; S glycoprotein
Host Species Rabbit
Immunogen Full-length SARS-CoV-2 Spike S1 protein
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage -20°C
Form Liquid
Isotype IgG
Show More Show Less

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.