Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FyterTech Nonwovens Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Click to view available options
Capacity (Metric):
0.94 L
3.78 L
18.9 L
Packaging Quantity:
1 Ea.
12/Cs.
4/Cs.
Description
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
- Sodium-free liquid reagent changes color when neutralization is complete
- Safe for use in laboratories and cleanrooms
Specifications
Specifications
Capacity (English) | 5 gal. |
Capacity (Metric) | 18.9 L |
Type | Base Neutralizer |
Format | Liquid |
For Use With (Application) | For lab and cleanrooms |
Includes | Color change indicator when neutralized |
Packaging Quantity | 1 Ea. |
Weight (English) | 50 lb. |
Weight (Metric) | 23 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction