Learn More
Labconco™ SpillStopper™ Work Surfaces for 5 Ft. Protector™ Premier™ and Protector™ XL Hoods
SDPSCI_ED
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector Premier and XL Hoods.
$2704.00
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work
View More Specs
Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S28230
SDPSCI_ED
View Documents
Labconco™
9500500/EMD
No Cutouts
Each of 1 for $2,704.00
Only null left
Add to Cart
Description
Description
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
Protector XL Laboratory Hood
Work Surface
Epoxy Resin
Laboratory Fume Hood Work
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.