Learn More
Neutec Group Spin Air Microbial Air Samplers
SDP
Catalog No. 14910435
Click to view available options
Description:
Spin Air Microbial Air Sampler
Spin Air Microbial Air Sampler Daughter Unit
Used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications
Microbial air sampling slave unit must be connected to a main Spin Air unit. When connected to a main unit it allows for duplicate sampling.
  • Rotating plate holder for even surface spread of micro organisms (Except the Basic Air model)
  • Sampling by volume (not timed) for higher accuracy and reproducibility
  • Utilizes standard 60mm or 90mm agar plates which are easy to find and inexpensive
  • Rechargeable battery operated for remote sampling
  • Delayed activation mode to avoid collector's contamination contribution
  • Optional duplicate simultaneous sampling with low cost daughter unit (Spin Air model only)
  • Bar code connectivity for faster easier collection of sample, location and user information (Spin Air Model)
  • Wide range of adaptors and accessories plastic and aluminum choices in 60mm, 90mm and Rodac sizes
Specifications
Description Spin Air Microbial Air Sampler
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.