Learn More
Invitrogen™ SPIN1 Polyclonal Antibody

Description
SPIN1 Polyclonal Antibody detects endogenous levels of SPIN1 protein.
Specifications
Specifications
| Antigen | SPIN1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SPIN1 |
| Gene Accession No. | Q4V8J7, Q61142, Q9Y657 |
| Gene Alias | 30000 Mr metaphase complex; OCR; Ovarian cancer-related protein; SPIN; Spin1; spindlin 1; Spindlin1; spindlin-1; SSEC P; TDRD24 |
| Gene Symbols | SPIN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.