Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599099 Supplier Invitrogen™ Supplier No. PA599099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPIN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SPIN1 (spindlin 1), also known as SPIN or ovarian cancer-related protein (OCR), is a 262 amino acid nuclear protein suggested to play a role in regulation of the cell-cycle during the transition from gamete to embryo. A member of the SPIN/STSY family, SPIN1 localizes to interphase nucleus and mitotic chromosomes, and is modified by phosphorylation in a cell-cycle-dependent fashion. A meiotic spindle-binding protein, SPIN1 overexpression has been shown to cause defects in mitotic spindles, thereby resulting in chromosome instability and potential tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIN1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPIN1
Gene Accession No. Q9Y657
Gene Alias 30000 Mr metaphase complex; OCR; Ovarian cancer-related protein; SPIN; Spin1; spindlin 1; Spindlin1; spindlin-1; SSEC P; TDRD24
Gene Symbols SPIN1
Host Species Rabbit
Immunogen Recombinant Human Spindlin-1 protein (1-262AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10927
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.