Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPIN2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570756
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Pig: 93%; Rabbit: 92%; Rat: 93%.
May be involved in the regulation of cell cycle progression.
Specifications
SPIN2A | |
Polyclonal | |
Unconjugated | |
SPIN2A | |
centromeric spin2 copy; dJ323P24.1; DXF34; Protein DXF34; RGD1560898; SPIN2; SPIN-2; SPIN2A; spindlin family member 2A; spindlin family, member 2; spindlin family, member 2A; spindlin homolog 2; spindlin-2; spindlin-2A; spindlin-like protein 2; spindlin-like protein 2A; TDRD25 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54466 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99865 | |
SPIN2A | |
synthetic peptide directed towards the N-terminal region of human SPIN2A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction