Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPIN2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567079 Supplier Invitrogen™ Supplier No. PA567079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKTPNAQEAEG QQTRAAAGRA TGSANMTKKK VSQ Highest antigen sequence identity to the following orthologs - mouse 53%, rat 53%.

May be involved in the regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIN2A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPIN2A
Gene Accession No. Q99865
Gene Alias centromeric spin2 copy; dJ323P24.1; DXF34; Protein DXF34; RGD1560898; SPIN2; SPIN-2; SPIN2A; spindlin family member 2A; spindlin family, member 2; spindlin family, member 2A; spindlin homolog 2; spindlin-2; spindlin-2A; spindlin-like protein 2; spindlin-like protein 2A; TDRD25
Gene Symbols SPIN2A
Host Species Rabbit
Immunogen Recombinant Human SPIN2A. Recombinant protein control fragment (Product #RP-105657).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54466
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.