Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPIN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550265 Supplier Invitrogen™ Supplier No. PA550265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SPIN4 protein.

Exhibits H3K4me3-binding activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIN4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SPIN4
Gene Accession No. Q56A73
Gene Alias 9630042H07Rik; FLJ44984; MGC133224; novel Spin/Ssty repeat containing protein; Spin4; spindlin family member 4; spindlin family, member 4; spindlin-4; TDRD28
Gene Symbols SPIN4
Host Species Rabbit
Immunogen Full length fusion protein of Human SPIN4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139886
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.