Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPIN90 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572616
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2616 10 μL
50-157-2615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2616 Supplier Bethyl Laboratories, Inc Supplier No. IHC00731T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Guinea pig_10141, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Chinese hamster and

The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintece of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified.

Specifications

Antigen SPIN90
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NCKIPSD
Gene Accession No. Q9NZQ3
Gene Alias AF3P21, DIP, DIP1, ORF1, SPIN90, WASLBP, WISH, 54 kDa VacA-interacting protein, 54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, DIP-1, NCK-interacting protein with SH3 domain, SH3 adapter protein SPIN90, SH3 protein interacting with Nck, 90 kDa, VIP54, WASP-interacting SH3-domain
Gene Symbols NCKIPSD
Host Species Rabbit
Immunogen Between 600 and 650
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51517
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.