Learn More
Stoelting™ Spinal Adaptor and Clamps
SDP
Holds mice or rats for spinal surgery
$1208.00 - $4020.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 4
Catalog Number Mfr. No. Model Animal Price Quantity & Availability  
Catalog Number Mfr. No. Model Animal Price Quantity & Availability  
10-000-184
SDP
View Documents
Stoelting™
51690
Spinal Adaptor Mouse
Each for $4,020.00
Only null left
Add to Cart
10-000-185
SDP
View Documents
Stoelting™
51691
Transverse Clamps, Set of 4 Mouse
Pack of 4 for $1,233.00
Only null left
Add to Cart
10-000-186
SDP
View Documents
Stoelting™
51692
Spinal Adaptor Rat
Pack of 4 for $1,208.00
Only null left
Add to Cart
10-000-187
SDP
View Documents
Stoelting™
51695
Transverse Clamps, Set of 4 Rat
Each for $3,950.00
Only null left
Add to Cart
Description
Description
  • Design inspired by the need for a versatile instrument for stabilizing the spinal column
  • Accessories to be mounted on the 51600, 51900 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.
  • Utilizes the manipulator and base plate of the stereotaxic instrument, enabling precision spinal preparations
  • Spinal column is secured by up to four independent transverse process clamps, as well as a spinous process clamp
  • Transverse process clamps are designed to minimize trauma while providing motionless support
  • Engineered for one-handed control of gross and fine adjustment of the surgical preparation in the AP, ML, and DV planes.
  • Transverse process clamps are available to accommodate either the rat or the mouse
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.