Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Spinal Needles

Catalog No. p-188275
Click to view available options
Needle Gauge:
18 G
20 G
22 G
Color:
Black
Pink
Yellow
Quantity:
10/Pk.
25/Pk.
Length (English):
3 in.
3.5 in.
5 in.
6 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

  • Key/Slot arrangement of stylet and cannula hubs facilitates proper needle
  • bevel orientation
  • Unique translucent window hub helps to allow for easy visualization of CSF
  • Fitted stylet reduces tissue coring

TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Color Pink
Needle Gauge 18 G
Includes Key/slot arrangement of fitted stylet, cannula hubs, translucent window hub
Latex Free Yes
Length (English) 3.5 in.
Product Type Spinal Needle
Disposable Single-use
Quantity 25/Pk.
Needle Point Style Bevel
Sterility Sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.