Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ Host Depletion Microbial DNA Kit

Catalog No. MP116545050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116545050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116545050 Supplier MP Biomedicals™ Supplier No. 116545050
Only null left
Add to Cart
Add to Cart

Provides an easy-to-use workflow to isolate microbial DNA from samples containing high amounts of host DNA. Selective lysis of host cells and depletion followed by microbial DNA purification. Lysing Matrix E for thorough and complete microbial lysis with FastPrep instruments.

  • The SPINeasy Host Depletion Microbial DNA Kit provides an easy-to-use workflow to isolate and enrich microbial DNA from samples containing high amounts of host DNA. Host DNA contamination is common in samples such as swabs and bodily fluids. The presence of high amounts of human DNA can limit the application and sensitivity of downstream shotgun metagenomic sequencing or microbiome analysis, as most of the extracted DNA is from human and not microbial origin.
  • The SPINeasy Host Depletion Microbial DNA Kit provides efficient reduction of host DNA from these samples by selective lysis of host cells with our specially formulated Host Lysis Buffer. Host DNA, which has been released into the solution, is then removed from the supernatant through centrifugation. A further host depletion treatment provides enzymatic degradation of any remaining host DNA, as well as DNA coming from dead microbial cells. Finally, the remaining intact microbial cells are lysed through a combination of specially formulated buffers and highly efficient homogenization using FastPrep instruments from MP Biomedicals. Enriched microbial DNA is then purified using a convenient silica membrane spin-column technology workflow and ready for downstream molecular applications, such as PCR and NGS.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Isolation Technology Spin Column
Sample Type Saliva, Swabs, Bodily Fluid
Includes 12 mL Equilibration buffer,50x Lysing Matrix E, 60 mL Host Lysis Buffer, 6 mL Host Depletion Buffer, 1 Vial Host Depletion Enzyme HDE, 200 μL HDE Reconstitution Buffer, 12 mL Microbial Selection Buffer, 40 mL Microbial Lysis Buffer, 30 mL Wash Buffer HD1, 6 mL Wash Buffer HD2, 6 mL Wash Buffer HD, 50x Column HD with collection tubes
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.