Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spinesin Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100896
Description
Spinesin Polyclonal specifically detects Spinesin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Spinesin | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, MGC141886, MGC148044, Spinesin, transmembrane protease serine 5, transmembrane protease, serine 5 | |
Goat | |
Affinity Purified | |
RUO | |
80975 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.5 - 1.5 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_110397 | |
TMPRSS5 | |
Peptide with sequence C-LDWIHDTAQDSLL corresponding to C-Terminus according to NP_110397.2. | |
0.1 mg | |
Primary | |
Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction