Learn More
Invitrogen™ Spinesin Monoclonal Antibody (376620)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Spinesin |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 376620 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | TMPRSS5 |
| Gene Accession No. | Q9H3S3 |
| Gene Alias | adrenal mitochondrial protease; Amp; MGC141886; MGC148044; Spinesin; Tmprss5; Transmembrane protease serine 5; transmembrane protease, serine 5; transmembrane protease, serine 5 (spinesin) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.