Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588061 Supplier Invitrogen™ Supplier No. PA588061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSLGREAKCY NELNGCTKIY DPVCGTDGNT YPNECVLCFE NRKRQTSILI QKSGPC; Positive Samples: Mouse intestine.

This is a trypsin inhibitor, its physiological function is to prevent the trypsin-catalyzed premature activation of zymogens within the pancreas. Defects in SPINK1 are a cause of pancreatitis (PCTT) [MIM:167800]. A disease characterized by the presence of calculi in pancreatic ducts. It causes severe abdominal pain attacks. Defects in SPINK1 are the cause of susceptibility to tropical calcific pancreatitis (TCP) [MIM:608189]. TCP is an idiopathic, juvenile, nonalcoholic form of chronic pancreatitis widely prevalent in several tropical countries. It can be associated with fibrocalculous pancreatic diabetes (FCPD) depending on both environmental and genetic factors. TCP differs from alcoholic pancreatitis by a much younger age of onset, pancreatic calcification, a high incidence of insulin dependent but ketosis resistant diabetes mellitus, and an exceptionally high incidence of pancreatic cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPINK1
Gene Accession No. P09036
Gene Alias calcium transport inhibitor; Caltrin; ISK1; P12; Pancreatic secretory trypsin inhibitor; pancreatic secretory trypsin inhibitor II; pancreatic secretory trypsin inhibitor type II (PSTI-II); PCTT; prostatic secretory glycoprotein; PSTI; PSTI-II; serine peptidase inhibitor Kazal type 1; serine peptidase inhibitor, Kazal type 1; serine peptidase inhibitor, Kazal type 3; serine protease inhibitor Kazal-type 1; Serine protease inhibitor Kazal-type 1-like; Serine protease inhibitor Kazal-type 3; serine protease inhibitor, Kazal type 1; serine protease inhibitor, Kazal type 3; Spink 3; Spink1; Spink1l; Spink3; TATI; TCP; Tumor-associated trypsin inhibitor
Gene Symbols SPINK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-79 of human SPINK1 (NP_0031132).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20730
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.