Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557668 Supplier Invitrogen™ Supplier No. PA557668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FNKRDFTRWP KPRCKMYIPL DPDYNADCPN VTAPVCASNG HTFQNECFFC VEQREFHYRI Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 53%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINK13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPINK13
Gene Accession No. Q1W4C9
Gene Alias HBVDNAPTP1; Hepatitis B Virus (HBV) DNA polymerase transactivated protein 1; hepatitis B virus DNA polymerase transactivated human serine protease inhibitor; hepatitis B virus DNA polymerase transactivated serine protease inhibitor; Hespintor; LiESP6; serine peptidase inhibitor, Kazal type 13 (putative); serine protease inhibitor Kazal-type 13; Serine protease inhibitor Kazal-type 5-like 3; SPINK13; SPINK5L3
Gene Symbols SPINK13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPINK13. Recombinant protein control fragment (Product #RP-96993).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.