Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555426 Supplier Invitrogen™ Supplier No. PA555426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KYRTPNCSQY RLPGCPRHFN PVCGSDMSTY ANECTLCMKI REGGHNIKII RNGPC Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 58%.

Strong inhibitor of acrosin in male and/or female genital tract. Also inhibits trypsin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINK2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPINK2
Gene Accession No. P20155
Gene Alias 1700007F22Rik; acrosin-trypsin inhibitor; AV038945; Epididymis tissue protein Li 172; HUSI-II; serine peptidase inhibitor, Kazal type 2; serine peptidase inhibitor, Kazal type 2 (acrosin-trypsin inhibitor); serine protease inhibitor Kazal-type 2; SPINK2
Gene Symbols SPINK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPINK2. Recombinant protein control fragment (Product #RP-89223).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.