Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK4 Antibody (248), DyLight 680 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289953FR
Description
SPINK4 Monoclonal antibody specifically detects SPINK4 in Human samples. It is validated for ELISASpecifications
SPINK4 | |
Monoclonal | |
DyLight 680 | |
gastrointestinal peptide, MGC133107, serine peptidase inhibitor, Kazal type 4, serine protease inhibitor Kazal-type 4, serine protease inhibitor, Kazal type 4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPINK4 (NP_055286.1; Met1-Cys86). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
248 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
27290 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction