Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552414 Supplier Invitrogen™ Supplier No. PA552414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPFSRMPICE HMVESPTCSQ MSNLVCGTDG LTYTNECQLC LARIKTKQDI QIMKDGK Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 65%.

SPINK4 (Serine Peptidase Inhibitor, Kazal Type 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include serine-type endopeptidase inhibitor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINK4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPINK4
Gene Accession No. O60575
Gene Alias epididymis luminal protein 136; gastrointestinal peptide; HEL136; PEC60; PEC-60; Peptide PEC-60 homolog; serine peptidase inhibitor, Kazal type 4; serine protease inhibitor Kazal-type 4; serine protease inhibitor, Kazal type 4; SPINK4
Gene Symbols SPINK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPINK4. Recombinant protein control fragment (Product #RP-89257).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27290
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.