Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK4 Recombinant Rabbit Monoclonal Antibody (265), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$293.50 - $565.00
Specifications
Antigen | SPINK4 |
---|---|
Clone | 265 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543353
|
Invitrogen™
MA543353 |
50 μL |
Each for $293.50
|
|
|||||
PIMA543142
![]() |
Invitrogen™
MA543142 |
100 μL |
Each for $565.00
|
|
|||||
Description
Specificity: Human SPINK4
SPINK4 (Serine Peptidase Inhibitor, Kazal Type 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include serine-type endopeptidase inhibitor activity.Specifications
SPINK4 | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
epididymis luminal protein 136; gastrointestinal peptide; HEL136; PEC60; PEC-60; Peptide PEC-60 homolog; serine peptidase inhibitor, Kazal type 4; serine protease inhibitor Kazal-type 4; serine protease inhibitor, Kazal type 4; SPINK4 | |
SPINK4 | |
IgG | |
Protein A | |
Antibody |
265 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
O60575 | |
27290 | |
Recombinant Human SPINK4 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
SPINK4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title