Learn More
Invitrogen™ SPINK5 Monoclonal Antibody (C10), Invitrogen™
Description
Specifications
Specifications
| Antigen | SPINK5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | C10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | SPINK5 |
| Gene Accession No. | Q9NQ38 |
| Gene Alias | 2310065D10Rik; AV238912; Hemofiltrate peptide HF6478; Hemofiltrate peptide HF7665; ISK5; LEKT1; LEKTI; LETK1; LETKI; lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; NETS; NS; serine peptidase inhibitor, Kazal type 5; serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5; SPINK5; VAKT1; VAKTI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.