Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPINK5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537458
Description
This antibody detects endogenous protein at a molecular weight of 130 kDa. Purity is >95% by SDS-PAGE.
Lympho-epithelial Kazal-type inhibitor (LEKTI) is a serine protease inhibitor which protects mucous epithelia against microbial attack and inflammation. LEKTI is a marker of epithelial differentiation and expresses strongly in the granular and uppermost spinous layers of the epidermis and differentiated layers of stratified epithelia. Defects in SPINK5, the gene encoding LEKTI, are the cause of Netherton syndrome, a severe autosomal recessive disorder characterized by atopic dermatitis, hayfever and other conditions.
Specifications
| SPINK5 | |
| Polyclonal | |
| Unconjugated | |
| SPINK5 | |
| 2310065D10Rik; AV238912; Hemofiltrate peptide HF6478; Hemofiltrate peptide HF7665; ISK5; LEKT1; LEKTI; LETK1; LETKI; lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; NETS; NS; serine peptidase inhibitor, Kazal type 5; serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5; SPINK5; VAKT1; VAKTI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11005 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQ38 | |
| SPINK5 | |
| Synthetic peptide corresponding to amino acids 496-543 of human LEKTI. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction