Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPINK6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550127
Description
The antibody detects endogenous levels of total SPINK6 protein.
SPINK6 (serine peptidase inhibitor, Kazal type 6), also known as BUSI2, is an 80 amino acid secreted protein that contains one kazal-like domain and is thought to function as a serine protease inhibitor. The gene encoding SPINK6 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also associated with chromosome 5 and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute mye-logenous leukemias and myelodysplastic syndrome.
Specifications
SPINK6 | |
Polyclonal | |
Unconjugated | |
SPINK6 | |
acrosin inhibitor; BUSI2; EG433180; Kallikrein inhibitor; Kazal type serine protease inhibitor 1; protease inhibitor H; serine peptidase inhibitor, Kazal type 6; serine protease inhibitor Kazal-type 6; Spink5l1; Spink6; UNQ844; UNQ844/PRO1782 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
404203 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6UWN8 | |
SPINK6 | |
Synthesized peptide derived from internal of human SPINK6 (aa 13-62). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction