Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA552779

Catalog No. PIPA552779


Only null left
Add to Cart

Description

Description

Immunogen sequence: VDCGEFQDTK VYCTRESNPH CGSDGQTYGN KCAFCKAIVK SGGKISLKH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

SPINK6 (serine peptidase inhibitor, Kazal type 6), also known as BUSI2, is an 80 amino acid secreted protein that contains one kazal-like domain and is thought to function as a serine protease inhibitor. The gene encoding SPINK6 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also associated with chromosome 5 and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute mye-logenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPINK6
Polyclonal
Unconjugated
SPINK6
acrosin inhibitor; BUSI2; EG433180; Kallikrein inhibitor; Kazal type serine protease inhibitor 1; protease inhibitor H; serine peptidase inhibitor, Kazal type 6; serine protease inhibitor Kazal-type 6; Spink5l1; Spink6; UNQ844; UNQ844/PRO1782
Rabbit
Antigen Affinity Chromatography
RUO
404203
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6UWN8
SPINK6
Recombinant protein corresponding to Human SPINK6. Recombinant protein control fragment (Product #RP-90497).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.