Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK7 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241141
Description
SPINK7 Polyclonal specifically detects SPINK7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SPINK7 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SPINK7 | |
Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ECRG2. The immunogen is located within the last 50 amino acids of ECRG2. Amino Acid Squence: TESLKSNGRVQFLHD | |
Affinity Purified | |
RUO | |
84651 | |
Human, Mouse, Rat | |
IgY |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
ECG2MGC133106, ECRG2, ECRG-2, esophagus cancer related gene 2, Esophagus cancer-related gene 2 protein, esophagus cancer-related gene-2, MGC133105, serine peptidase inhibitor, Kazal type 7 (putative), serine protease inhibitor Kazal-type 7 | |
Chicken | |
14 kDa | |
0.1 mg | |
Primary | |
ECRG2 antibody is predicted to not cross-react with other ECRG family members | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction